Assay Details
Gene Symbol: Cdipt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026638.3 | NP_080914.1 | 4 - 5 | 1262 | 81 | |
GenBank mRNA | AK076974.1 | - | 4 - 5 | 1262 | 81 | |
AK079226.1 | - | 4 - 5 | 739 | 81 | ||
AK089109.1 | - | 4 - 5 | 828 | 81 | ||
BC021473.1 | - | 4 - 5 | 790 | 5343411 | 81 | |
BC024413.1 | - | 4 - 5 | 602 | 4973307 | 81 | |
BC032182.1 | - | 4 - 5 | 799 | 5252470 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
lipid metabolic process
phosphatidylinositol biosynthetic process
phospholipid biosynthetic process
CDP-diacylglycerol metabolic process
CDP-diacylglycerol-inositol 3-phosphatidyltransferase activity
transferase activity
phosphotransferase activity, for other substituted phosphate groups
diacylglycerol binding
manganese ion binding
carbohydrate binding
alcohol binding
lipid metabolic process
phosphatidylinositol biosynthetic process
phospholipid biosynthetic process
CDP-diacylglycerol metabolic process
CDP-diacylglycerol-inositol 3-phosphatidyltransferase activity
transferase activity
phosphotransferase activity, for other substituted phosphate groups
diacylglycerol binding
manganese ion binding
carbohydrate binding
alcohol binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503357 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503358 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503356 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503359 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503354 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503355 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408471 | BC032182.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408471 | AK132286.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408471 | AK207162.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408471 | AK214056.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408471 | BC021473.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408471 | BC032182.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408471 | AK132286.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408471 | AK207162.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408471 | AK214056.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408471 | BC021473.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 404306 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 217028 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 48046 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655625 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155556 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408471 | BC032182.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408471 | AK132286.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408471 | AK207162.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408471 | AK214056.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408471 | BC021473.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408471 | BC032182.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408471 | AK132286.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408471 | AK207162.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408471 | AK214056.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408471 | BC021473.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P408471 | BC032182.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P408471 | AK132286.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P408471 | AK207162.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P408471 | AK214056.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P408471 | BC021473.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P408471 | BC032182.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P408471 | AK132286.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P408471 | AK207162.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P408471 | AK214056.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P408471 | BC021473.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503370 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503343 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503320 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503357 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503350 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503332 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503323 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503361 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503341 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503362 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503333 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503336 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503322 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503368 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503328 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503338 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503352 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503360 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503349 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503324 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503354 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503345 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503330 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503318 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503327 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503366 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503363 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503335 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503365 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503355 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503353 | - | - |
Whole Mouse Genome G4122A | A_51_P408471 | BC032182.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P408471 | AK132286.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P408471 | AK207162.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P408471 | AK214056.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P408471 | BC021473.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P408471 | BC032182.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P408471 | AK132286.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P408471 | AK207162.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P408471 | AK214056.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P408471 | BC021473.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121511 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06478665 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06036838 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06478664 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06478673 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06187443 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06187444 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06478675 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06478668 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503358 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503359 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503354 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503356 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503357 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503317 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503350 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503343 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503341 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503391 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06647354 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503400 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503350 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503368 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503366 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503377 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503338 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503370 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503317 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503323 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503392 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503345 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503387 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503341 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503325 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503383 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05906121 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06375845 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503393 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06381043 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06187444 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503399 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06285888 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503355 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503361 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503389 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06201142 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503379 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503369 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503390 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06285889 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503373 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06264387 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503365 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503343 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06528283 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06405640 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06478665 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503360 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06528281 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06405639 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06595542 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06285895 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503320 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503375 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503353 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06375837 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503330 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503314 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503335 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06187436 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503349 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06099816 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25503395 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10557562 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10557565 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10557570 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10557568 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10557563 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10557567 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10557569 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10557564 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10557561 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1436715_PM_s_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1423657_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10557560 | - | - |
Mouse Genome 430 2.0 Array | 1436715_s_at | - | - |
Mouse Genome 430 2.0 Array | 1436715_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1423657_at | - | - |
Mouse Genome 430 2.0 Array | 1423657_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52858_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52858_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700015689.mm.1 | TC0700001827.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700015689.mm.1 | NM_026638.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700015691.mm.1 | TC0700001827.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700015691.mm.1 | NM_026638.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700015688.mm.1 | TC0700001827.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700015692.mm.1 | TC0700001827.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700015692.mm.1 | NM_026638.3 | Refseq |
Affymetrix HT Mouse Genome 430A Array | 1436715_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1436715_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1423657_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1423657_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6110706 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006110706 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1224421 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1224421 | - | - |
mouse-6 v1.1 (Illumina) | scl32044.6_195-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6110706 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl32044.6_195-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6110706 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1224421 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3101385 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3101385 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1224421 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl32044.6_195-S | - | - |
