Assay Details
Gene Symbol: Cdc26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139291.3 | NP_647452.1 | 2 - 3 | 167 | 73 | |
GenBank mRNA | AK012627.1 | - | 2 - 3 | 168 | 73 | |
AK041840.1 | - | 1 - 2 | 1057 | 73 | ||
AK047055.1 | - | 187 | 73 | |||
AK135556.1 | - | 168 | 73 | |||
BC005775.1 | - | 1 - 2 | 94 | 3602527 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK012627.1,AK135556.1,BC005775.1,NM_139291.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22964276 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22964275 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139112 | AK135556.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139112 | BC005775.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139112 | BC114350.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139112 | AK086692.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139112 | AK047055.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 255372 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 38766 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 322132 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 427684 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 681377 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 552291 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 135472 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 210050 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669760 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554551 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 682890 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 948598 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 114509 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12311 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139112 | AK135556.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139112 | BC005775.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139112 | BC114350.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139112 | AK086692.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139112 | AK047055.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P649285 | AK047055.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P649285 | AK008199.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P649285 | AK047055.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P649285 | AK008199.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P161526 | AK041840.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353663 | AK135556.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353663 | AK047055.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353663 | AK028893.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353663 | AK034911.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P1180267 | AK135556.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P1180267 | AK028893.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964274 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964255 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964269 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964256 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964272 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964267 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964268 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964254 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964261 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964260 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964259 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964276 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964263 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964273 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05257333 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05257306 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05257325 | - | - |
Whole Mouse Genome G4122A | A_52_P50959 | AK135556.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P50959 | BC005775.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P50959 | BC114350.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P50959 | AK086692.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P50959 | AK047055.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P353663 | AK135556.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P353663 | AK047055.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P353663 | AK028893.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P353663 | AK034911.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P161526 | AK041840.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P1180267 | AK135556.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P1180267 | AK028893.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136141 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05257325 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03720525 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03524316 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03720521 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03720514 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03522693 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03522696 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03522692 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03720504 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22964276 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22964275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964230 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964256 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964204 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964212 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964239 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22964247 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22964259 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22964241 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03445411 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22964253 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03874196 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22964267 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22964264 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22964239 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03989474 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22964250 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22964261 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03290688 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22964272 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22964262 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22964246 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22964274 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22964263 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03720504 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22964238 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22964260 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22964243 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03522696 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22964237 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10416270 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10513585 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10513586 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10513584 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1452992_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1456860_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1423987_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10416269 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10513583 | - | - |
Mouse Genome 430 2.0 Array | 1452992_at | - | - |
Mouse Genome 430 2.0 Array | 1456860_at | - | - |
Mouse Genome 430 2.0 Array | 1423987_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1423987_at | - | - |
Mouse Genome 430 2.0 Array | 1452992_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1456860_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66440_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66440_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0400022855.mm.1 | TC0400002787.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400022855.mm.1 | NM_139291.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400022855.mm.1 | NM_139291.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400022856.mm.1 | TC0400002787.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400022856.mm.1 | NM_139291.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400022856.mm.1 | NM_139291.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400022849.mm.1 | TC0400002787.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400022849.mm.1 | NM_139291.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400022849.mm.1 | NM_139291.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400022852.mm.1 | TC0400002787.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400022852.mm.1 | NM_139291.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400022852.mm.1 | NM_139291.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400022851.mm.1 | TC0400002787.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400022851.mm.1 | NM_139291.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400022851.mm.1 | NM_139291.3 | Refseq |
Mouse Expression Set 430 | 1423987_at | - | - |
Mouse Expression Set 430 | 1423987_PM_at | - | - |
Mouse Expression Set 430 | 1439974_at | - | - |
Mouse Expression Set 430 | 1456860_at | - | - |
Mouse Expression Set 430 | 1441486_PM_at | - | - |
Mouse Expression Set 430 | 1439974_PM_at | - | - |
Mouse Expression Set 430 | 1452992_PM_at | - | - |
Mouse Expression Set 430 | 1452992_at | - | - |
Mouse Expression Set 430 | 1441486_at | - | - |
Mouse Expression Set 430 | 1456860_PM_at | - | - |
Murine Genome U74v2 Set | 94084_at | - | - |
Murine Genome U74v2 Set | 94084_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1423987_at | - | - |
Mouse Genome 430A 2.0 Array | 1423987_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1452992_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1452992_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456860_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456860_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 344932 | - | - |
ABI Mouse Genome Survey Microarray | 383575 | - | - |
ABI Mouse Genome Survey Microarray | 620426 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 101410110 | - | - |
MouseWG-6 v2 Expression BeadChip | 5570279 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 106620136 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5570279 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 101410110 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005570279 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2588986 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2588986 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2445936 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2506519 | - | - |
mouse-6 v1.1 (Illumina) | scl25379.1.1_72-S | - | - |
mouse-6 v1.1 (Illumina) | scl0066440.2_150-S | - | - |
mouse-6 v1.1 (Illumina) | scl24266.6.1_112-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 101410110 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5570279 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 106620136 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0066440.2_150-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5570279 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101410110 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106620136 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2588986 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2445936 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2588986 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0066440.2_150-S | - | - |
