Assay Details
| Gene Symbol: Mettl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144877.1 | NP_659126.1 | 5 - 6 | 1561 | 63 | |
| GenBank mRNA | AK084750.1 | - | 5 - 6 | 1256 | 63 | |
| AK146751.1 | - | 1759 | 63 | |||
| AK168959.1 | - | 5 - 6 | 1768 | 63 | ||
| AK169319.1 | - | 1760 | 63 | |||
| AK173044.1 | - | 1714 | 63 | |||
| BC014872.1 | - | 5 - 6 | 1561 | 3602469 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mettl13
Genomic Map