Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600002461.mm.1 |
TC0600000331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002461.mm.1 |
NM_145575.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002461.mm.1 |
NM_145575.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002458.mm.1 |
TC0600000331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002458.mm.1 |
NM_145575.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002458.mm.1 |
NM_145575.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002460.mm.1 |
TC0600000331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002460.mm.1 |
NM_145575.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002460.mm.1 |
NM_145575.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002463.mm.1 |
TC0600000331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002463.mm.1 |
NM_145575.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002463.mm.1 |
NM_145575.3 |
Refseq |