Assay Details
| Gene Symbol: Msantd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145609.1 | NP_663584.1 | 1 - 2 | 623 | 101 | |
| GenBank mRNA | AK032884.1 | - | 2 - 3 | 674 | 101 | |
| AK132023.1 | - | 2 - 3 | 659 | 101 | ||
| BC014729.1 | - | 1 - 2 | 623 | 4461077 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2099695 | AK132023.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2099695 | BC014729.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2099695 | NM_145609.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 239140 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 457090 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 2911 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P131969 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127212 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2099695 | AK132023.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2099695 | BC014729.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2099695 | NM_145609.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276698 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276706 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276709 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276715 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276702 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276720 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276716 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276722 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26276712 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127212 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06688155 | - | - |
| Whole Mouse Genome G4122A | A_51_P108269 | AK132023.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P108269 | BC014729.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P108269 | NM_145609.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P108269 | AK032884.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106862 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127212 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06688173 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07645646 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07645635 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08103043 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08081968 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07645641 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08081971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26276687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08012480 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07729527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07749574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07852698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07622295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07749578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276720 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08186579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07729531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08012479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08125606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07606991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276653 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08125604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26276690 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1424993_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424993_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424993_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78100_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78100_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900000209.mm.1 | NM_145609.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900000209.mm.1 | NM_145609.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900000209.mm.1 | TC0900000026.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900000208.mm.1 | NM_145609.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900000208.mm.1 | NM_145609.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900000208.mm.1 | TC0900000026.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900000207.mm.1 | NM_145609.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900000207.mm.1 | NM_145609.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900000207.mm.1 | TC0900000026.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424993_PM_at | - | - |
| Mouse Expression Set 430 | 1424993_at | - | - |
| Murine Genome U74v2 Set | 138978_at | - | - |
| Murine Genome U74v2 Set | 138978_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424993_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424993_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 432033 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6520025 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006520025 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1232530 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1232530 | - | - |
| mouse-6 v1.1 (Illumina) | scl37319.3_438-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6520025 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37319.3_438-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6520025 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1232530 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1232530 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl37319.3_438-S | - | - |

Msantd4
Genomic Map