Assay Details
Gene Symbol: Ranbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145824.4 | NP_665823.2 | 12 - 13 | 1787 | 119 | |
GenBank mRNA | AK087165.1 | - | 2 - 3 | 204 | 119 | |
AK173177.1 | - | 12 - 13 | 1600 | 119 | ||
AY337314.1 | - | 12 - 13 | 1788 | 119 | ||
BC024698.1 | - | 12 - 13 | 1632 | 3667000 | 119 | |
EU281316.1 | - | 12 - 13 | 1648 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
