Assay Details
Gene Symbol: Cd37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290802.1 | NP_001277731.1 | 4 - 5 | 432 | 58 | |
NM_001290804.1 | NP_001277733.1 | 3 - 4 | 357 | 58 | ||
NM_007645.4 | NP_031671.1 | 3 - 4 | 381 | 58 | ||
GenBank mRNA | AK154250.1 | - | 3 - 4 | 420 | 58 | |
AK154466.1 | - | 4 - 5 | 402 | 58 | ||
AK155663.1 | - | 3 - 4 | 358 | 58 | ||
AK155966.1 | - | 3 - 4 | 346 | 58 | ||
BC019402.1 | - | 3 - 4 | 418 | 4013070 | 58 | |
BE849269.1 | - | 3 - 4 | 299 | 58 | ||
CT010217.1 | - | 3 - 4 | 270 | 58 | ||
CX226822.1 | - | 4 - 5 | 433 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
