Assay Details
| Gene Symbol: Cd37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290802.1 | NP_001277731.1 | 8 - 9 | 934 | 74 | |
| NM_001290804.1 | NP_001277733.1 | 7 - 8 | 859 | 74 | ||
| NM_007645.4 | NP_031671.1 | 7 - 8 | 883 | 74 | ||
| GenBank mRNA | AK154466.1 | - | 8 - 9 | 904 | 74 | |
| AK155966.1 | - | 7 - 8 | 848 | 74 | ||
| BC019402.1 | - | 7 - 8 | 920 | 4013070 | 74 | |
| BY548149.1 | - | 1 - 2 | 95 | 74 | ||
| CT010217.1 | - | 7 - 8 | 772 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25091980 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25092029 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25091983 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25092034 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25092028 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25091981 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25092033 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25091982 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25092032 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25092031 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25092030 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25091984 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2174490 | CT010217.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2174490 | AK155663.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2174490 | AK155966.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2174490 | BC019402.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2174490 | NM_007645.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2118570 | BC019402.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2118570 | NM_007645.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 760515 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 249327 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655166 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165981 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2118570 | BC019402.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2118570 | NM_007645.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2174490 | CT010217.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2174490 | AK155663.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2174490 | AK155966.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2174490 | BC019402.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2174490 | NM_007645.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | ERCC-00040_174 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P350752 | BC019402.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092037 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092030 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165981 | - | - |
| Whole Mouse Genome G4122A | A_52_P463373 | CT010217.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P463373 | AK155663.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P463373 | AK155966.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P463373 | BC019402.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P463373 | NM_007645.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P350752 | BC019402.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105775 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165981 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06353384 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06340697 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06340696 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06340699 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06340698 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06353383 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25091980 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25092031 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25091983 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25091984 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25092028 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25092032 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25092029 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25092030 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25091981 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25092034 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25091982 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25092033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092027 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06608876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06666207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06340697 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06666211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06666214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092033 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06397551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06397563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092027 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06506216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06340699 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06608875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092015 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10563182 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563189 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563186 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563185 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563184 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563183 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563188 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563179 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563181 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563180 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563187 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419206_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425736_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10563178 | - | - |
| Mouse Genome 430 2.0 Array | 1419206_at | - | - |
| Mouse Genome 430 2.0 Array | 1425736_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1425736_at | - | - |
| Mouse Genome 430 2.0 Array | 1419206_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12493_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12493_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700027039.mm.1 | NM_007645.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700027039.mm.1 | NM_007645.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700027039.mm.1 | TC0700002999.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700027040.mm.1 | NM_007645.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700027040.mm.1 | NM_007645.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700027040.mm.1 | TC0700002999.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419206_PM_at | - | - |
| Mouse Expression Set 430 | 1425736_at | - | - |
| Mouse Expression Set 430 | 1425736_PM_at | - | - |
| Mouse Expression Set 430 | 1419206_at | - | - |
| Murine Genome U74v2 Set | 98980_at | - | - |
| Murine Genome U74v2 Set | 98980_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425736_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419206_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425736_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419206_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425736_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419206_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425736_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419206_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 770273 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000770273 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1228126 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1228126 | - | - |
| mouse-6 v1.1 (Illumina) | scl31384.10.152_39-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 770273 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31384.10.152_39-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 770273 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2929526 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2929526 | - | - |

Cd37
Genomic Map