Assay Details
| Gene Symbol: Cd53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007651.3 | NP_031677.1 | 6 - 7 | 700 | 69 | |
| GenBank mRNA | AK156510.1 | - | 6 - 7 | 607 | 69 | |
| AK160326.1 | - | 6 - 7 | 598 | 69 | ||
| BC021310.1 | - | 6 - 7 | 614 | 5010267 | 69 | |
| BC052905.1 | - | 6 - 7 | 565 | 30078163 | 69 | |
| X97227.1 | - | 5 - 6 | 501 | 69 | ||
| Z16071.1 | - | 5 - 6 | 486 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd53
Genomic Map