Assay Details
| Gene Symbol: Cd53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007651.3 | NP_031677.1 | 7 - 8 | 784 | 97 | |
| GenBank mRNA | AK156510.1 | - | 7 - 8 | 691 | 97 | |
| AK160326.1 | - | 7 - 8 | 682 | 97 | ||
| BC021310.1 | - | 7 - 8 | 698 | 5010267 | 97 | |
| BC052905.1 | - | 7 - 8 | 649 | 30078163 | 97 | |
| X97227.1 | - | 6 - 7 | 585 | 97 | ||
| Z16071.1 | - | 6 - 7 | 570 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P337675 | X97227.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P337675 | Z16071.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P337675 | AK156510.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P337675 | AK160326.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P337675 | BC021310.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P337675 | X97227.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P337675 | Z16071.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P337675 | AK156510.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P337675 | AK160326.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P337675 | BC021310.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 890003 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 353093 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 439041 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576779 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 140893 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 469924 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 280127 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 403994 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 345311 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 802219 | BC051930.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 802219 | NM_026000.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 653342 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 373174 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 805020 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 6814 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 875902 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 561020 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 793744 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494632 | BC031493.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494632 | XM_001477340.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494632 | XM_001478143.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 866332 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 24380 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 135352 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 37348 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 121833 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174806 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102936 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P337675 | X97227.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P337675 | Z16071.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P337675 | AK156510.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P337675 | AK160326.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P337675 | BC021310.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P337675 | X97227.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P337675 | Z16071.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P337675 | AK156510.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P337675 | AK160326.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P337675 | BC021310.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402874 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05020992 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174806 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05021020 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102936 | - | - |
| Whole Mouse Genome G4122A | A_51_P337675 | X97227.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P337675 | Z16071.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P337675 | AK156510.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P337675 | AK160326.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P337675 | BC021310.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P337675 | X97227.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P337675 | Z16071.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P337675 | AK156510.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P337675 | AK160326.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P337675 | BC021310.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116839 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174806 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102936 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02306138 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02306120 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02306136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02306138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02974215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02306120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02306144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02306131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402847 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402848 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02306124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02306139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02453200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02306142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02306127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02469568 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10501065 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501070 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501066 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501071 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501069 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501067 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501064 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501068 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1459744_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448617_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10501063 | - | - |
| Mouse Genome 430 2.0 Array | 1439589_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1459744_at | - | - |
| Mouse Genome 430 2.0 Array | 1459744_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448617_at | - | - |
| Mouse Genome 430 2.0 Array | 1448617_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439589_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12508_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12508_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300020992.mm.1 | NM_007651.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300020992.mm.1 | NM_007651.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300020992.mm.1 | TC0300002695.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300020994.mm.1 | NM_007651.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300020994.mm.1 | NM_007651.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300020994.mm.1 | TC0300002695.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448617_PM_at | - | - |
| Mouse Expression Set 430 | 1448617_at | - | - |
| Mouse Expression Set 430 | 1459744_at | - | - |
| Mouse Expression Set 430 | 1459744_PM_at | - | - |
| Murine Genome U74v2 Set | 94939_at | - | - |
| Murine Genome U74v2 Set | 94939_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448617_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448617_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448617_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448617_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459744_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459744_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5550162 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5550162 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105420722 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005550162 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2657478 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2657478 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2481559 | - | - |
| mouse-6 v1.1 (Illumina) | scl0012508.2_215-S | - | - |
| mouse-6 v1.1 (Illumina) | scl21690.8_397-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105420722 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5550162 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0012508.2_215-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5550162 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105420722 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2657478 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2481559 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2657478 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0012508.2_215-S | - | - |

Cd53
Genomic Map