Assay Details
| Gene Symbol: Cd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007657.3 | NP_031683.1 | 5 - 6 | 558 | 64 | |
| GenBank mRNA | AK002251.1 | - | 5 - 6 | 564 | 64 | |
| AK012793.1 | - | 5 - 6 | 564 | 64 | ||
| AK151443.1 | - | 5 - 6 | 524 | 64 | ||
| AK151619.1 | - | 5 - 6 | 511 | 64 | ||
| AK170310.1 | - | 5 - 6 | 511 | 64 | ||
| BC070474.1 | - | 5 - 6 | 558 | 30634238 | 64 | |
| L08115.1 | - | 5 - 6 | 496 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd9
Genomic Map