Assay Details
| Gene Symbol: Dnm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253893.1 | NP_001240822.1 | 10 - 11 | 1525 | 118 | |
| NM_001253894.1 | NP_001240823.1 | 10 - 11 | 1525 | 118 | ||
| NM_007871.2 | NP_031897.2 | 10 - 11 | 1525 | 118 | ||
| GenBank mRNA | AK005012.2 | - | 10 - 11 | 1501 | 118 | |
| AK154375.1 | - | 9 - 10 | 1189 | 118 | ||
| AK172231.1 | - | 10 - 11 | 1506 | 118 | ||
| L31398.1 | - | 10 - 11 | 1465 | 118 | ||
More Information
Set Membership: |
|

Dnm2
Genomic Map