Assay Details
| Gene Symbol: Dpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007876.2 | NP_031902.2 | 4 - 5 | 626 | 84 | |
| GenBank mRNA | AK002380.1 | - | 3 - 4 | 506 | 84 | |
| BC003492.1 | - | 4 - 5 | 496 | 2812088 | 84 | |
| D13139.1 | - | 4 - 5 | 84 | |||
More Information
Set Membership: |
|

Dpep1
Genomic Map