Assay Details
| Gene Symbol: Dpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007876.2 | NP_031902.2 | 5 - 6 | 782 | 64 | |
| GenBank mRNA | BC003492.1 | - | 5 - 6 | 652 | 2812088 | 64 |
| D13139.1 | - | 5 - 6 | 782 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dpep1
Genomic Map