Assay Details
| Gene Symbol: Ecm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252653.1 | NP_001239582.1 | 1 - 2 | 195 | 66 | |
| NM_007899.2 | NP_031925.2 | 1 - 2 | 195 | 66 | ||
| GenBank mRNA | AK153009.1 | - | 1 - 2 | 194 | 66 | |
| AK153471.1 | - | 1 - 2 | 194 | 66 | ||
| AK159334.1 | - | 1 - 2 | 194 | 66 | ||
| AK169939.1 | - | 1 - 2 | 197 | 66 | ||
| BC138693.1 | - | 1 - 2 | 181 | 8861715 | 66 | |
| BC145381.1 | - | 1 - 2 | 181 | 9053939 | 66 | |
| BY224903.1 | - | 1 - 2 | 196 | 66 | ||
| L33416.1 | - | 1 - 2 | 188 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ecm1
Genomic Map