Assay Details
Gene Symbol: Gnb2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008143.3 | NP_032169.1 | 4 - 5 | 628 | 79 | |
GenBank mRNA | AK002493.1 | - | 4 - 5 | 617 | 79 | |
AK012242.1 | - | 3 - 4 | 415 | 79 | ||
AK017772.1 | - | 4 - 5 | 629 | 79 | ||
AK051226.1 | - | 4 - 5 | 629 | 79 | ||
AK159797.1 | - | 4 - 5 | 629 | 79 | ||
AK160528.1 | - | 4 - 5 | 599 | 79 | ||
AK160739.1 | - | 4 - 5 | 626 | 79 | ||
AK161234.1 | - | 4 - 5 | 629 | 79 | ||
AK166417.1 | - | 4 - 5 | 617 | 79 | ||
AK166786.1 | - | 4 - 5 | 631 | 79 | ||
AK166800.1 | - | 4 - 5 | 632 | 79 | ||
AK166889.1 | - | 3 - 4 | 269 | 79 | ||
AK166945.1 | - | 4 - 5 | 629 | 79 | ||
AK168086.1 | - | 4 - 5 | 629 | 79 | ||
AK168096.1 | - | 4 - 5 | 629 | 79 | ||
AK168196.1 | - | 4 - 5 | 629 | 79 | ||
AK168349.1 | - | 4 - 5 | 660 | 79 | ||
AK169612.1 | - | 4 - 5 | 628 | 78 | ||
BC020322.1 | - | 4 - 5 | 605 | 79 | ||
BC036295.1 | - | 2 - 3 | 168 | 79 | ||
BC046760.1 | - | 4 - 5 | 613 | 5693613 | 79 | |
D29802.1 | - | 4 - 5 | 622 | 79 | ||
X75313.1 | - | 4 - 5 | 628 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
