Assay Details
Gene Symbol: Gss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291111.1 | NP_001278040.1 | 4 - 5 | 484 | 102 | |
NM_008180.2 | NP_032206.1 | 5 - 6 | 619 | 102 | ||
GenBank mRNA | AK146486.1 | - | 5 - 6 | 524 | 102 | |
AK149577.1 | - | 4 - 5 | 389 | 102 | ||
AK159630.1 | - | 5 - 6 | 544 | 102 | ||
AK169096.1 | - | 5 - 6 | 539 | 102 | ||
BC003784.1 | - | 5 - 6 | 532 | 3593913 | 102 | |
U35456.1 | - | 4 - 5 | 515 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
