Assay Details
| Gene Symbol: Hsd17b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008291.3 | NP_032317.2 | 2 - 3 | 259 | 93 | |
| GenBank mRNA | BB621153.1 | - | 2 - 3 | 259 | 93 | |
| U66827.1 | - | 2 - 3 | 244 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsd17b3
Genomic Map