Assay Details
Gene Symbol: Idua | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008325.4 | NP_032351.2 | 14 - 15 | 2246 | 67 | |
NM_172997.1 | NP_766585.1 | 13 - 14 | 2105 | 67 | ||
GenBank mRNA | AK031414.1 | - | 13 - 14 | 1900 | 67 | |
AK045330.1 | - | 12 - 13 | 1733 | 67 | ||
AK169998.1 | - | 13 - 14 | 1904 | 67 | ||
AK171124.1 | - | 9 - 10 | 1256 | 67 | ||
BC057935.1 | - | 13 - 14 | 1867 | 5327150 | 67 | |
L34111.1 | - | 13 - 14 | 1820 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis
carbohydrate metabolic process
lysosome organization
metabolic process
dermatan sulfate catabolic process
limb morphogenesis
skeletal system morphogenesis
chemical homeostasis
L-iduronidase activity
hydrolase activity, hydrolyzing O-glycosyl compounds
hydrolase activity
hydrolase activity, acting on glycosyl bonds
cell morphogenesis
carbohydrate metabolic process
lysosome organization
metabolic process
dermatan sulfate catabolic process
limb morphogenesis
skeletal system morphogenesis
chemical homeostasis
L-iduronidase activity
hydrolase activity, hydrolyzing O-glycosyl compounds
hydrolase activity
hydrolase activity, acting on glycosyl bonds
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23973434 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097178 | BQ175218.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097178 | NM_008325.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097176 | AK171124.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097176 | AK169998.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097176 | AK045330.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097176 | NM_008325.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 628619 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 402231 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 456282 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 907403 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 18465 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 283688 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385426 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 775221 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 324998 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546494 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 776120 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 48740 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169530 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129984 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126377 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097178 | BQ175218.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097178 | NM_008325.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097176 | AK171124.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097176 | AK169998.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097176 | AK045330.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097176 | NM_008325.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P114214 | L34111.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23973434 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23973436 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23973435 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P169530 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126377 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05690388 | - | - |
Whole Mouse Genome G4122A | A_51_P114214 | L34111.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110079 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126377 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P169530 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04271903 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04271955 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23973434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973435 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973551 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973442 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973432 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973564 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973428 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973608 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973582 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973402 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973575 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973415 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973444 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973515 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973500 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973541 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973572 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973440 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973409 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973559 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973411 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973591 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973588 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973495 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973430 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973422 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973571 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973577 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973436 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973583 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973561 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973580 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973573 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973509 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04271909 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04943152 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04271938 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23973420 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23973430 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04271898 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23973443 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04271916 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04271920 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04271921 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23973421 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23973448 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04271929 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23973436 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04271907 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23973419 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04271906 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23973432 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23973422 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23973445 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04271932 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04271939 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10524041 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10524038 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10524039 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10524042 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10524035 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10524051 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10524043 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10524040 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10524048 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10524049 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10524036 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10524044 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10524050 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10524045 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10524047 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10524037 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10524046 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418793_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1449230_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10532241 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10524034 | - | - |
Mouse Genome 430 2.0 Array | 1439251_at | - | - |
Mouse Genome 430 2.0 Array | 1418793_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1449230_at | - | - |
Mouse Genome 430 2.0 Array | 1449230_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1439251_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1418793_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15932_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15932_at | - | - |
Affymetrix Murine Genome 6500A Array | Msa13960_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0500009502.mm.1 | TC0500001112.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500009502.mm.1 | NM_008325.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500009502.mm.1 | NM_172997.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500009508.mm.1 | TC0500001112.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500009508.mm.1 | NM_008325.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500009508.mm.1 | NM_172997.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500009505.mm.1 | TC0500001112.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500009505.mm.1 | NM_008325.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500009505.mm.1 | NM_172997.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500009503.mm.1 | TC0500001112.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500009503.mm.1 | NM_008325.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500009503.mm.1 | NM_172997.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500009504.mm.1 | TC0500001112.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500009504.mm.1 | NM_008325.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500009504.mm.1 | NM_172997.1 | Refseq |
Mouse Expression Set 430 | 1418793_at | - | - |
Mouse Expression Set 430 | 1449230_PM_at | - | - |
Mouse Expression Set 430 | 1449230_at | - | - |
Mouse Expression Set 430 | 1418793_PM_at | - | - |
Murine Genome U74v2 Set | 104500_PM_at | - | - |
Murine Genome U74v2 Set | 104500_at | - | - |
Mouse Genome 430A 2.0 Array | 1418793_at | - | - |
Mouse Genome 430A 2.0 Array | 1449230_at | - | - |
Mouse Genome 430A 2.0 Array | 1418793_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1449230_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418793_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449230_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449230_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418793_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4060541 | - | - |
MouseWG-6 v2 Expression BeadChip | 2690131 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2690131 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4060541 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002690131 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004060541 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2676924 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2634213 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2676924 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2634213 | - | - |
mouse-6 v1.1 (Illumina) | scl0004080.1_2-S | - | - |
mouse-6 v1.1 (Illumina) | scl015932.14_0-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4060541 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2690131 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl015932.14_0-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0004080.1_2-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4060541 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2690131 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2676925 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2980466 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2676924 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2676925 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2980466 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2676924 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0004080.1_2-S | - | - |
