Assay Details
| Gene Symbol: Il13ra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK089687.2 | - | 4 - 5 | 844 | 93 | |
| U65747.1 | - | 1 - 2 | 108 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK089687.2, U65747.1 transcripts.
Set Membership: |
|

Il13ra2
Genomic Map