Assay Details
| Gene Symbol: Il13ra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK089687.2 | - | 5 - 6 | 953 | 98 | |
| BC003723.1 | - | 2 - 3 | 129 | 3486223 | 98 | |
| U65747.1 | - | 2 - 3 | 217 | 98 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK089687.2,BC003723.1,U65747.1 transcripts.
Set Membership: |
|

Il13ra2
Genomic Map