Assay Details
| Gene Symbol: Kif5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008448.3 | NP_032474.2 | 21 - 22 | 2785 | 66 | |
| GenBank mRNA | BC090841.1 | - | 21 - 22 | 2761 | 30543821 | 66 |
| L27153.1 | - | 21 - 22 | 2379 | 66 | ||
| U86090.1 | - | 21 - 22 | 2612 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kif5b
Genomic Map