Assay Details
| Gene Symbol: Car11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009800.4 | NP_033930.1 | 5 - 6 | 879 | 58 | |
| GenBank mRNA | AB079601.1 | - | 5 - 6 | 658 | 58 | |
| AF050105.1 | - | 3 - 4 | 432 | 58 | ||
| AK013465.1 | - | 5 - 6 | 879 | 58 | ||
| AY075026.1 | - | 5 - 6 | 880 | 58 | ||
| BC019393.1 | - | 5 - 6 | 864 | 3968862 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Car11
Genomic Map