Assay Details
| Gene Symbol: Clip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039162.2 | NP_001034251.1 | 3 - 4 | 1141 | 69 | |
| NM_009990.3 | NP_034120.2 | 3 - 4 | 1141 | 69 | ||
| GenBank mRNA | AK045967.1 | - | 3 - 4 | 985 | 69 | |
| AK172928.1 | - | 3 - 4 | 1034 | 69 | ||
| BC039162.1 | - | 3 - 4 | 1081 | 3498067 | 69 | |
| BC053048.1 | - | 3 - 4 | 1141 | 5709279 | 69 | |
| BC121191.1 | - | 3 - 4 | 787 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32982273 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32982271 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32982272 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P360112 | NM_001039162.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P360112 | NM_009990.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P360112 | AK045967.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P360112 | BC039162.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P360112 | BC053048.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P360112 | NM_001039162.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P360112 | NM_009990.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P360112 | AK045967.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P360112 | BC039162.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P360112 | BC053048.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 103212 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342973 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 117399 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 876677 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 27686 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 51566 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508459 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 458278 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 266314 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 694118 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 943587 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 207237 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551232 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 86713 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 262266 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546582 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 180677 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 120634 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 159084 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 298576 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375971 | AK006580.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 798264 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 845058 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923127 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115678 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 397053 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 311364 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 575369 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 249659 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736511 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740086 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 520648 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 480205 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 244441 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 404599 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506249 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 680691 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724984 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412314 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 234189 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 289294 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 663295 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 29022 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 614689 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 57625 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 106587 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 75518 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 57888 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 48079 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 9219 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P360112 | NM_001039162.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P360112 | NM_009990.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P360112 | AK045967.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P360112 | BC039162.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P360112 | BC053048.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P360112 | NM_001039162.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P360112 | NM_009990.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P360112 | AK045967.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P360112 | BC039162.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P360112 | BC053048.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P360112 | NM_001039162.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P360112 | NM_009990.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P360112 | AK045967.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P360112 | BC039162.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P360112 | BC053048.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P360112 | NM_001039162.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P360112 | NM_009990.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P360112 | AK045967.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P360112 | BC039162.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P360112 | BC053048.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P139296 | NM_001039162.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P139296 | NM_009990.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P139296 | AK045967.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P139296 | BC039162.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P139296 | BC053048.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P360112 | NM_001039162.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P360112 | NM_009990.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P360112 | AK045967.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P360112 | BC039162.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P360112 | BC053048.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P360112 | NM_001039162.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P360112 | NM_009990.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P360112 | AK045967.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P360112 | BC039162.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P360112 | BC053048.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139296 | NM_001039162.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139296 | NM_009990.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139296 | AK045967.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139296 | BC039162.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139296 | BC053048.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117416 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32982273 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32982272 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32982271 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32983175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982305 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982311 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982280 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982303 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982271 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982301 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32983178 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982277 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982281 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982268 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982255 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982225 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982283 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982247 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32983176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982309 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982276 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982278 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982312 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32982232 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10534293 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534295 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534286 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534289 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534297 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534285 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534284 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534298 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534287 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534292 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534283 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534288 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534299 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534300 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534291 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534282 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534294 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534296 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534290 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422984_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10534281 | - | - |
| Mouse Genome 430 2.0 Array | 1422984_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422984_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 269713_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 269713_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500030419.mm.1 | TC0500003373.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500030419.mm.1 | NM_001039162.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500030419.mm.1 | NM_009990.3 | Refseq |
| Mouse Expression Set 430 | 1422984_PM_at | - | - |
| Mouse Expression Set 430 | 1422984_at | - | - |
| Murine Genome U74v2 Set | 99432_at | - | - |
| Murine Genome U74v2 Set | 99432_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422984_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422984_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422984_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422984_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 862910 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3610010 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3610010 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1170148 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102650398 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001170148 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003610010 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1235107 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2698400 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2495703 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2698400 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1235107 | - | - |
| mouse-6 v1.1 (Illumina) | scl0004005.1_3-S | - | - |
| mouse-6 v1.1 (Illumina) | scl25944.17_344-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00269713.2_106-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102650398 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3610010 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1170148 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0004005.1_3-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00269713.2_106-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3610010 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1170148 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102650398 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2698400 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2630160 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2698400 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2495703 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2630160 | - | - |

Clip2
Genomic Map