Assay Details
| Gene Symbol: Ddt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010027.1 | NP_034157.1 | 2 - 3 | 336 | 92 | |
| GenBank mRNA | AK144193.1 | - | 2 - 3 | 379 | 92 | |
| BC010753.1 | - | 2 - 3 | 330 | 4241633 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P414706 | NM_010027.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P414706 | AK144193.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P414706 | BC010753.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P414706 | NM_010027.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P414706 | AK144193.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P414706 | BC010753.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 750733 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P120523 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P414706 | NM_010027.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P414706 | AK144193.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P414706 | BC010753.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P414706 | NM_010027.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P414706 | AK144193.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P414706 | BC010753.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258570 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258571 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P120523 | - | - |
| Whole Mouse Genome G4122A | A_51_P414706 | NM_010027.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P414706 | AK144193.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P414706 | BC010753.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P414706 | NM_010027.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P414706 | AK144193.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P414706 | BC010753.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130458 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P120523 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08315629 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08315627 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08629639 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08315610 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08629638 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08629640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08315618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258572 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10369991 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369992 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369990 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417103_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10369989 | - | - |
| Mouse Genome 430 2.0 Array | 1417103_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417103_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13202_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13202_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000017028.mm.1 | NM_010027.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000017028.mm.1 | NM_010027.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000017028.mm.1 | TC1000003225.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000017030.mm.1 | NM_010027.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000017030.mm.1 | NM_010027.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000017030.mm.1 | TC1000003225.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000024054.mm.1 | NM_010027.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000024054.mm.1 | NM_010027.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000024054.mm.1 | TC1000003225.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417103_PM_at | - | - |
| Mouse Expression Set 430 | 1417103_at | - | - |
| Murine Genome U74v2 Set | 100564_at | - | - |
| Murine Genome U74v2 Set | 100564_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417103_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417103_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417103_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417103_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 397332 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4590398 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4590398 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004590398 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2596297 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2596297 | - | - |
| mouse-6 v1.1 (Illumina) | scl37813.3.1_14-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4590398 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37813.3.1_14-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4590398 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2596297 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2596297 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl37813.3.1_14-S | - | - |

Ddt
Genomic Map