Assay Details
| Gene Symbol: Dhfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010049.3 | NP_034179.1 | 2 - 3 | 609 | 99 | |
| GenBank mRNA | AK031241.1 | - | 2 - 3 | 243 | 99 | |
| AK136212.1 | - | 2 - 3 | 667 | 99 | ||
| BC005796.1 | - | 2 - 3 | 181 | 3707582 | 99 | |
| BU058996.1 | - | 2 - 3 | 609 | 99 | ||
| L26316.1 | - | 2 - 3 | 188 | 99 | ||
| V00734.1 | - | 2 - 3 | 245 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dhfr
Genomic Map