Assay Details
| Gene Symbol: Faah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010173.4 | NP_034303.3 | 10 - 11 | 1520 | 91 | |
| GenBank mRNA | AK004985.1 | - | 10 - 11 | 1346 | 91 | |
| AK079598.1 | - | 10 - 11 | 1521 | 91 | ||
| AK080603.1 | - | 10 - 11 | 1353 | 91 | ||
| AK138481.1 | - | 10 - 11 | 1140 | 91 | ||
| BC006863.1 | - | 10 - 11 | 1299 | 3595191 | 91 | |
| BC052321.1 | - | 10 - 11 | 1562 | 6515634 | 91 | |
| U82536.1 | - | 10 - 11 | 1281 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Faah
Genomic Map