Assay Details
| Gene Symbol: Faah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010173.4 | NP_034303.3 | 12 - 13 | 1604 | 70 | |
| GenBank mRNA | AK004985.1 | - | 11 - 12 | 2128 | 70 | |
| AK079598.1 | - | 12 - 13 | 1605 | 70 | ||
| AK080603.1 | - | 12 - 13 | 1437 | 70 | ||
| AK138481.1 | - | 12 - 13 | 1224 | 70 | ||
| BC006863.1 | - | 12 - 13 | 1383 | 3595191 | 70 | |
| BC052321.1 | - | 12 - 13 | 1646 | 6515634 | 70 | |
| U82536.1 | - | 12 - 13 | 1365 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Faah
Genomic Map