Assay Details
| Gene Symbol: Gda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010266.2 | NP_034396.1 | 8 - 9 | 996 | 140 | |
| GenBank mRNA | AF174583.1 | - | 8 - 9 | 922 | 140 | |
| AF369880.1 | - | 8 - 9 | 881 | 140 | ||
| AF369881.1 | - | 8 - 9 | 881 | 140 | ||
| AF369882.1 | - | 8 - 9 | 881 | 140 | ||
| AK041056.1 | - | 8 - 9 | 935 | 140 | ||
| AK079419.1 | - | 8 - 9 | 804 | 140 | ||
| AK079459.1 | - | 8 - 9 | 936 | 140 | ||
| AK139617.1 | - | 764 | 140 | |||
| AK157014.1 | - | 8 - 9 | 958 | 140 | ||
| AK173138.1 | - | 8 - 9 | 925 | 140 | ||
| BC076583.1 | - | 8 - 9 | 909 | 30620457 | 140 | |
More Information
Set Membership: |
|

Gda
Genomic Map