Assay Details
| Gene Symbol: Gda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010266.2 | NP_034396.1 | 11 - 12 | 1303 | 104 | |
| GenBank mRNA | AF174583.1 | - | 11 - 12 | 1229 | 104 | |
| AF369880.1 | - | 11 - 12 | 1188 | 104 | ||
| AF369881.1 | - | 11 - 12 | 1188 | 104 | ||
| AF369882.1 | - | 11 - 12 | 1188 | 104 | ||
| AF369883.1 | - | 11 - 12 | 1188 | 104 | ||
| AF369884.1 | - | 11 - 12 | 1188 | 104 | ||
| AK041056.1 | - | 11 - 12 | 1242 | 104 | ||
| AK079419.1 | - | 11 - 12 | 1111 | 104 | ||
| AK079459.1 | - | 11 - 12 | 1242 | 104 | ||
| AK139617.1 | - | 1072 | 104 | |||
| AK157014.1 | - | 11 - 12 | 1265 | 104 | ||
| AK173138.1 | - | 11 - 12 | 1232 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gda
Genomic Map