Assay Details
| Gene Symbol: Gnb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010313.2 | NP_034443.1 | 3 - 4 | 340 | 85 | |
| NM_138719.5 | NP_619733.1 | 1 - 2 | 316 | 85 | ||
| GenBank mRNA | AK050564.1 | - | 1 - 2 | 221 | 85 | |
| AK148175.1 | - | 1 - 2 | 211 | 85 | ||
| AK149908.1 | - | 1 - 2 | 133 | 85 | ||
| AK169323.1 | - | 1 - 2 | 249 | 85 | ||
| BC016135.1 | - | 3 - 4 | 274 | 4506201 | 85 | |
| CK621143.1 | - | 3 - 4 | 340 | 85 | ||
| L34290.1 | - | 1 - 2 | 142 | 85 | ||
| U69145.1 | - | 2 - 3 | 238 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gnb5
Genomic Map