Assay Details
| Gene Symbol: Gnb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010313.2 | NP_034443.1 | 7 - 8 | 726 | 92 | |
| NM_138719.5 | NP_619733.1 | 5 - 6 | 702 | 92 | ||
| NR_073186.1 | 6 - 7 | 896 | 92 | |||
| GenBank mRNA | AK018747.1 | - | 3 - 4 | 256 | 92 | |
| AK050564.1 | - | 5 - 6 | 607 | 92 | ||
| AK148175.1 | - | 5 - 6 | 597 | 92 | ||
| AK149908.1 | - | 5 - 6 | 519 | 92 | ||
| AK169323.1 | - | 5 - 6 | 635 | 92 | ||
| BC016135.1 | - | 7 - 8 | 660 | 4506201 | 92 | |
| L34290.1 | - | 5 - 6 | 528 | 92 | ||
| U69145.1 | - | 6 - 7 | 624 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gnb5
Genomic Map