Assay Details
| Gene Symbol: Gnb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010313.2 | NP_034443.1 | 8 - 9 | 874 | 60 | |
| NM_138719.5 | NP_619733.1 | 6 - 7 | 850 | 60 | ||
| NR_073186.1 | 7 - 8 | 1044 | 60 | |||
| GenBank mRNA | AK018747.1 | - | 4 - 5 | 404 | 60 | |
| AK050564.1 | - | 6 - 7 | 755 | 60 | ||
| AK148175.1 | - | 6 - 7 | 745 | 60 | ||
| AK169323.1 | - | 6 - 7 | 783 | 60 | ||
| AK195270.1 | - | 1 - 2 | 56 | 60 | ||
| AK198237.1 | - | 1 - 2 | 56 | 60 | ||
| AK214864.1 | - | 1 - 2 | 56 | 60 | ||
| BC016135.1 | - | 8 - 9 | 808 | 4506201 | 60 | |
| L34290.1 | - | 6 - 7 | 676 | 60 | ||
| U69145.1 | - | 7 - 8 | 772 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gnb5
Genomic Map