Assay Details
| Gene Symbol: Gnb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010313.2 | NP_034443.1 | 9 - 10 | 968 | 79 | |
| NM_138719.5 | NP_619733.1 | 7 - 8 | 944 | 79 | ||
| NR_073186.1 | 8 - 9 | 1138 | 79 | |||
| GenBank mRNA | AK018747.1 | - | 5 - 6 | 498 | 79 | |
| AK050564.1 | - | 7 - 8 | 849 | 79 | ||
| AK148175.1 | - | 7 - 8 | 839 | 79 | ||
| AK149908.1 | - | 7 - 8 | 761 | 79 | ||
| AK169323.1 | - | 7 - 8 | 877 | 79 | ||
| AK189847.1 | - | 2 - 3 | 150 | 79 | ||
| AK197902.1 | - | 2 - 3 | 150 | 79 | ||
| AK198237.1 | - | 2 - 3 | 150 | 79 | ||
| AK214864.1 | - | 2 - 3 | 150 | 79 | ||
| BC016135.1 | - | 9 - 10 | 902 | 4506201 | 79 | |
| L34290.1 | - | 7 - 8 | 770 | 79 | ||
| U69145.1 | - | 8 - 9 | 866 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gnb5
Genomic Map