Assay Details
| Gene Symbol: Hdac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010411.2 | NP_034541.2 | 13 - 14 | 1134 | 87 | |
| GenBank mRNA | AF074881.1 | - | 13 - 14 | 1081 | 87 | |
| AF074882.1 | - | 7 - 8 | 508 | 87 | ||
| AF098295.1 | - | 13 - 14 | 1059 | 87 | ||
| AF125536.1 | - | 13 - 14 | 1066 | 87 | ||
| AK145158.1 | - | 13 - 14 | 1100 | 87 | ||
| BC139300.1 | - | 13 - 14 | 1162 | 8862322 | 87 | |
| BC139301.1 | - | 13 - 14 | 1162 | 8862323 | 87 | |
| JN651901.1 | - | 7 - 8 | 782 | 87 | ||
| JN651902.1 | - | 6 - 7 | 740 | 87 | ||
| JN651903.1 | - | 7 - 8 | 491 | 87 | ||
| JN651904.1 | - | 4 - 5 | 336 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hdac3
Genomic Map