Assay Details
| Gene Symbol: Hmox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010442.2 | NP_034572.1 | 3 - 4 | 767 | 86 | |
| GenBank mRNA | AK150934.1 | - | 3 - 4 | 764 | 86 | |
| AK151005.1 | - | 3 - 4 | 764 | 86 | ||
| AK151012.1 | - | 3 - 4 | 756 | 86 | ||
| AK151627.1 | - | 3 - 4 | 743 | 86 | ||
| AK153420.1 | - | 3 - 4 | 764 | 86 | ||
| AK153516.1 | - | 3 - 4 | 764 | 86 | ||
| AK153566.1 | - | 3 - 4 | 764 | 86 | ||
| AK154598.1 | - | 3 - 4 | 765 | 86 | ||
| AK159521.1 | - | 3 - 4 | 764 | 86 | ||
| AK169227.1 | - | 3 - 4 | 764 | 86 | ||
| BC010757.1 | - | 3 - 4 | 736 | 4235374 | 86 | |
| M33203.1 | - | 3 - 4 | 709 | 86 | ||
| X13356.1 | - | 3 - 4 | 709 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hmox1
Genomic Map