Assay Details
| Gene Symbol: Idh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111320.1 | NP_001104790.1 | 3 - 4 | 604 | 64 | |
| NM_010497.3 | NP_034627.3 | 3 - 4 | 582 | 64 | ||
| GenBank mRNA | AF020039.1 | - | 2 - 3 | 432 | 64 | |
| AK087063.1 | - | 3 - 4 | 484 | 64 | ||
| AK149019.1 | - | 3 - 4 | 513 | 64 | ||
| AK151212.1 | - | 3 - 4 | 507 | 64 | ||
| AK159173.1 | - | 3 - 4 | 513 | 64 | ||
| AK160896.1 | - | 3 - 4 | 514 | 64 | ||
| AK167158.1 | - | 3 - 4 | 554 | 64 | ||
| AK167583.1 | - | 3 - 4 | 518 | 64 | ||
| AK168832.1 | - | 3 - 4 | 556 | 64 | ||
| BC088986.1 | - | 3 - 4 | 554 | 6808731 | 64 | |
| CF950262.1 | - | 3 - 4 | 543 | 64 | ||
| DQ403075.1 | - | 2 - 3 | 332 | 64 | ||
More Information
Set Membership: |
|

Idh1
Genomic Map