Assay Details
| Gene Symbol: Il18rap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010553.4 | NP_034683.1 | 1 - 2 | 122 | 137 | |
| GenBank mRNA | AF077347.1 | - | 1 - 2 | 131 | 137 | |
| AK137541.1 | - | 1 - 2 | 83 | 137 | ||
| BC120598.1 | - | 1 - 2 | 129 | 40129521 | 137 | |
| BC120600.1 | - | 1 - 2 | 129 | 40129523 | 137 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF077347.1,AK137541.1,BC120598.1,BC120600.1,NM_010553.4 transcripts.
Set Membership: |
|

Il18rap
Genomic Map