Assay Details
| Gene Symbol: Cnot7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271542.1 | NP_001258471.1 | 3 - 4 | 547 | 132 | |
| NM_001271543.1 | NP_001258472.1 | 3 - 4 | 618 | 132 | ||
| NM_011135.5 | NP_035265.1 | 3 - 4 | 618 | 132 | ||
| GenBank mRNA | AK009561.1 | - | 3 - 4 | 606 | 133 | |
| AK044535.1 | - | 2 - 3 | 1484 | 132 | ||
| AK132063.1 | - | 3 - 4 | 538 | 132 | ||
| AK135669.1 | - | 3 - 4 | 549 | 132 | ||
| AK137838.1 | - | 3 - 4 | 583 | 132 | ||
| AK147048.1 | - | 3 - 4 | 585 | 132 | ||
| AK166449.1 | - | 3 - 4 | 582 | 132 | ||
| BC006021.1 | - | 3 - 4 | 562 | 3590764 | 132 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cnot7
Genomic Map