Assay Details
| Gene Symbol: Farsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278075.1 | NP_001265004.1 | 3 - 4 | 323 | 91 | |
| NM_011811.4 | NP_035941.2 | 3 - 4 | 323 | 91 | ||
| NR_102757.1 | 3 - 4 | 225 | 91 | |||
| NR_102758.1 | 2 - 3 | 267 | 91 | |||
| GenBank mRNA | AF123263.1 | - | 3 - 4 | 323 | 91 | |
| AK010271.1 | - | 3 - 4 | 286 | 91 | ||
| AK012154.1 | - | 3 - 4 | 286 | 91 | ||
| AK045432.1 | - | 3 - 4 | 275 | 91 | ||
| AK076607.1 | - | 3 - 4 | 285 | 91 | ||
| AK134799.1 | - | 2 - 3 | 229 | 91 | ||
| AK165645.1 | - | 3 - 4 | 190 | 91 | ||
| AK167945.1 | - | 3 - 4 | 286 | 91 | ||
| AK168923.1 | - | 3 - 4 | 267 | 91 | ||
| AK169621.1 | - | 2 - 3 | 230 | 91 | ||
| BC002313.1 | - | 3 - 4 | 182 | 3592805 | 91 | |
| BC016428.1 | - | 3 - 4 | 290 | 4239906 | 91 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK134799.1 transcripts.
Set Membership: |
|

Farsb
Genomic Map