Assay Details
| Gene Symbol: Farsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278075.1 | NP_001265004.1 | 12 - 13 | 1220 | 84 | |
| NM_011811.4 | NP_035941.2 | 12 - 13 | 1220 | 84 | ||
| NR_102757.1 | 12 - 13 | 1122 | 84 | |||
| NR_102758.1 | 11 - 12 | 1164 | 84 | |||
| GenBank mRNA | AF123263.1 | - | 12 - 13 | 1220 | 84 | |
| AK010271.1 | - | 12 - 13 | 1183 | 84 | ||
| AK012154.1 | - | 12 - 13 | 1183 | 84 | ||
| AK045432.1 | - | 12 - 13 | 1172 | 84 | ||
| AK076607.1 | - | 12 - 13 | 1182 | 84 | ||
| AK134799.1 | - | 11 - 12 | 1126 | 84 | ||
| AK165645.1 | - | 12 - 13 | 1087 | 84 | ||
| AK167945.1 | - | 12 - 13 | 1183 | 84 | ||
| AK168923.1 | - | 12 - 13 | 1164 | 84 | ||
| AK169621.1 | - | 11 - 12 | 1127 | 84 | ||
| BC002313.1 | - | 12 - 13 | 1079 | 3592805 | 84 | |
| BC016428.1 | - | 12 - 13 | 1187 | 4239906 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Farsb
Genomic Map