Assay Details
Gene Symbol: Irf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013674.1 | NP_038702.1 | 6 - 7 | 816 | 78 | |
GenBank mRNA | AK089319.1 | - | 6 - 7 | 835 | 78 | |
AK089819.1 | - | 6 - 7 | 838 | 78 | ||
AK154138.1 | - | 6 - 7 | 840 | 78 | ||
AK154315.1 | - | 6 - 7 | 828 | 78 | ||
AK156562.1 | - | 6 - 7 | 838 | 78 | ||
BC137713.1 | - | 6 - 7 | 804 | 8860732 | 78 | |
BC137714.1 | - | 6 - 7 | 804 | 8860733 | 78 | |
U11692.1 | - | 6 - 7 | 816 | 78 | ||
U34307.1 | - | 6 - 7 | 810 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
