Assay Details
| Gene Symbol: Casp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277932.1 | NP_001264861.1 | 2 - 3 | 779 | 60 | |
| NM_015733.5 | NP_056548.2 | 2 - 3 | 779 | 60 | ||
| GenBank mRNA | AB019600.1 | - | 2 - 3 | 536 | 60 | |
| AB019601.1 | - | 2 - 3 | 536 | 60 | ||
| AK085095.1 | - | 2 - 3 | 568 | 60 | ||
| AK085152.1 | - | 2 - 3 | 535 | 60 | ||
| AK154495.1 | - | 2 - 3 | 549 | 60 | ||
| AK156500.1 | - | 2 - 3 | 682 | 60 | ||
| AK162863.1 | - | 2 - 3 | 559 | 60 | ||
| BC056372.1 | - | 2 - 3 | 561 | 6834067 | 60 | |
| BC056447.1 | - | 2 - 3 | 779 | 5721188 | 60 | |
| CT010400.1 | - | 2 - 3 | 536 | 60 | ||
More Information
Set Membership: |
|

Casp9
Genomic Map