Assay Details
| Gene Symbol: Enpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136077.2 | NP_001129549.1 | 3 - 4 | 476 | 72 | |
| NM_001285994.1 | NP_001272923.1 | 3 - 4 | 476 | 72 | ||
| NM_001285995.1 | NP_001272924.1 | 3 - 4 | 476 | 72 | ||
| NM_015744.3 | NP_056559.2 | 3 - 4 | 476 | 72 | ||
| GenBank mRNA | AF123542.1 | - | 3 - 4 | 332 | 72 | |
| AK038940.1 | - | 3 - 4 | 456 | 72 | ||
| AK088491.1 | - | 3 - 4 | 349 | 72 | ||
| AK161144.1 | - | 3 - 4 | 349 | 72 | ||
| AK163881.1 | - | 3 - 4 | 349 | 72 | ||
| BC003264.1 | - | 3 - 4 | 353 | 3499038 | 72 | |
| BC058759.1 | - | 3 - 4 | 439 | 6491897 | 72 | |
| EU131009.1 | - | 3 - 4 | 291 | 72 | ||
| EU131010.1 | - | 3 - 4 | 291 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Enpp2
Genomic Map