| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400019994.mm.1 |
NM_001136056.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019994.mm.1 |
TC0400002475.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400019992.mm.1 |
NM_001136056.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019992.mm.1 |
NM_016673.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019992.mm.1 |
NM_016673.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019992.mm.1 |
TC0400002475.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400019993.mm.1 |
NM_001136056.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019993.mm.1 |
NM_016673.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019993.mm.1 |
NM_016673.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019993.mm.1 |
TC0400002475.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400019989.mm.1 |
TC0400002475.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400019991.mm.1 |
NM_001136056.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019991.mm.1 |
NM_016673.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019991.mm.1 |
NM_016673.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019991.mm.1 |
TC0400002475.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400019990.mm.1 |
NM_001136056.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019990.mm.1 |
NM_016673.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019990.mm.1 |
NM_016673.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019990.mm.1 |
TC0400002475.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400019999.mm.1 |
NM_016673.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019999.mm.1 |
NM_016673.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019999.mm.1 |
TC0400002475.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400019998.mm.1 |
NM_016673.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019998.mm.1 |
NM_016673.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019998.mm.1 |
TC0400002475.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400019995.mm.1 |
NM_001136056.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019995.mm.1 |
TC0400002475.mm.1 |
MTA TC |