Assay Details
| Gene Symbol: Cntfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136056.2 | NP_001129528.1 | 5 - 6 | 663 | 76 | |
| NM_016673.2 | NP_057882.2 | 5 - 6 | 642 | 76 | ||
| GenBank mRNA | AB154246.1 | - | 3 - 4 | 438 | 76 | |
| AF068615.1 | - | 5 - 6 | 593 | 76 | ||
| AK137026.1 | - | 5 - 6 | 552 | 76 | ||
| BC046974.1 | - | 5 - 6 | 632 | 6389656 | 76 | |
| BC050928.1 | - | 5 - 6 | 660 | 6415187 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cntfr
Genomic Map