Assay Details
| Gene Symbol: Cntfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136056.2 | NP_001129528.1 | 8 - 9 | 1180 | 98 | |
| NM_016673.2 | NP_057882.2 | 8 - 9 | 1159 | 98 | ||
| GenBank mRNA | AF068615.1 | - | 8 - 9 | 1110 | 98 | |
| AK137026.1 | - | 8 - 9 | 1069 | 98 | ||
| BC046974.1 | - | 8 - 9 | 1149 | 6389656 | 98 | |
| BC050928.1 | - | 8 - 9 | 1177 | 6415187 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cntfr
Genomic Map