Assay Details
Gene Symbol: Gal3st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177691.1 | NP_001171162.1 | 2 - 3 | 212 | 103 | |
NM_001177703.1 | NP_001171174.1 | 2 - 3 | 343 | 103 | ||
NM_016922.3 | NP_058618.2 | 3 - 4 | 561 | 103 | ||
GenBank mRNA | AB032939.1 | - | 1 - 2 | 138 | 103 | |
AK002510.1 | - | 2 - 3 | 212 | 103 | ||
AK007645.1 | - | 2 - 3 | 343 | 103 | ||
BC026806.1 | - | 3 - 4 | 484 | 4215616 | 103 | |
CX239940.1 | - | 2 - 3 | 255 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27555125 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27555132 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27555126 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013823 | BC026806.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013823 | AK002510.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013823 | AK007645.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013823 | AB032939.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013823 | BB804195.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 357005 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 900808 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 512135 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 895750 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 390985 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 94398 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 29948 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 514979 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 237782 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 140184 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 45824 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117247 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114896 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013823 | BC026806.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013823 | AK002510.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013823 | AK007645.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013823 | AB032939.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013823 | BB804195.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P354354 | BC026806.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P354354 | AK002510.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P354354 | AB032939.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P354354 | NM_001177691.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P354354 | NM_001177703.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555023 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555018 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555044 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555038 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555035 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555048 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555047 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555027 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555041 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555031 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555053 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555050 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555028 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555051 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555037 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555033 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555045 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555024 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555026 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117247 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114896 | - | - |
Whole Mouse Genome G4122A | A_51_P354354 | BC026806.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P354354 | AK002510.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P354354 | AB032939.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P354354 | NM_001177691.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P354354 | NM_001177703.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133451 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114896 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117247 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09416690 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09416684 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09416686 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09416688 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09403403 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09416681 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27555132 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27555125 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27555126 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555117 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555098 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555149 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555094 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555171 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555066 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555060 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555175 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555059 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555184 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555061 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555136 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555139 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555098 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555085 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09514462 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555075 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09074247 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555079 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555047 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555115 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555091 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555035 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09011046 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555061 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555101 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09011053 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555055 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555086 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555122 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555051 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555050 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555044 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555018 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09011051 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555100 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555070 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555063 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555074 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555083 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09518063 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09507864 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555066 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555080 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555048 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555057 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555120 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09685822 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555112 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555093 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09403406 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555117 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09507870 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555072 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555108 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555028 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555031 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555053 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555021 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555026 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555056 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555107 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09742375 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09685825 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09418936 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555088 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555089 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09507869 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555041 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555023 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555064 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555024 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555059 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555038 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555078 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10373831 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10373829 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10373830 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10373828 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1454078_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10373826 | - | - |
Mouse Genome 430 2.0 Array | 1454078_a_at | - | - |
Mouse Genome 430 2.0 Array | 1454078_PM_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 53897_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 53897_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100000362.mm.1 | NM_001177691.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000362.mm.1 | NM_001177691.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000362.mm.1 | NM_001177703.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000362.mm.1 | NM_001177703.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000362.mm.1 | NM_016922.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000362.mm.1 | NM_016922.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000362.mm.1 | TC1100000029.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100000359.mm.1 | NM_001177703.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000359.mm.1 | NM_001177703.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000359.mm.1 | NM_016922.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000359.mm.1 | NM_016922.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000359.mm.1 | TC1100000029.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100000361.mm.1 | NM_001177691.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000361.mm.1 | NM_001177691.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000361.mm.1 | TC1100000029.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100000360.mm.1 | NM_016922.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000360.mm.1 | NM_016922.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000360.mm.1 | TC1100000029.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100000363.mm.1 | NM_001177691.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000363.mm.1 | NM_001177691.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000363.mm.1 | NM_001177703.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000363.mm.1 | NM_001177703.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000363.mm.1 | NM_016922.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000363.mm.1 | NM_016922.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000363.mm.1 | TC1100000029.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100000358.mm.1 | NM_001177703.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000358.mm.1 | NM_001177703.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000358.mm.1 | NM_016922.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000358.mm.1 | NM_016922.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000358.mm.1 | TC1100000029.mm.1 | MTA TC |
Mouse Expression Set 430 | 1454078_PM_a_at | - | - |
Mouse Expression Set 430 | 1454078_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1454078_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1454078_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1454078_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1454078_PM_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 610073 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5080746 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5080746 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005080746 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1258028 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1258028 | - | - |
mouse-6 v1.1 (Illumina) | scl41896.5_609-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5080746 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl41896.5_609-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5080746 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1258028 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1258028 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl41896.5_609-S | - | - |
