Assay Details
| Gene Symbol: Klrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016970.1 | NP_058666.1 | 3 - 4 | 380 | 81 | |
| GenBank mRNA | AF097357.1 | - | 3 - 4 | 380 | 81 | |
| AJ010751.1 | - | 3 - 4 | 429 | 81 | ||
| BC103546.1 | - | 3 - 4 | 379 | 40044831 | 81 | |
| BC103547.1 | - | 3 - 4 | 379 | 40044830 | 81 | |
| BC103548.1 | - | 3 - 4 | 379 | 40044829 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988844 | NM_016970.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988844 | AF097357.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 156483 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 129369 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 372556 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 134540 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 386154 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 609644 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 148092 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179482 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140508 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145031 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988844 | NM_016970.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988844 | AF097357.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P256983 | BC103546.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P256983 | BC103547.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P256983 | BC101953.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P256983 | BC103548.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P256983 | NM_016970.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24807852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24807859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24807884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24807868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24807869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24807882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24807865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24807880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24807872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24807864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24807862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24807874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24807879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24807867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24807855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24807851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24807860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24807877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24807861 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179482 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140508 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145031 | - | - |
| Whole Mouse Genome G4122A | A_51_P256983 | BC103546.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P256983 | BC103547.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P256983 | BC101953.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P256983 | BC103548.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P256983 | NM_016970.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125898 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120653 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140508 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179482 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145031 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05662913 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05662902 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05662904 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05662901 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05662892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24807763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807821 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05715506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05662908 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05041423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807880 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807836 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807843 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05662888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807832 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807826 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05662896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05342961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807879 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05077519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807892 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807884 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05662909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807857 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807819 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807816 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05077517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05878343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05662893 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24807841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05404295 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10547594 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547596 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547591 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547593 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547595 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547592 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420788_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10547590 | - | - |
| Mouse Genome 430 2.0 Array | 1420788_at | - | - |
| Mouse Genome 430 2.0 Array | 1420788_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50928_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50928_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600023682.mm.1 | TC0600003062.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600023682.mm.1 | NM_016970.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600023682.mm.1 | NM_016970.1 | Refseq |
| Mouse Expression Set 430 | 1420788_at | - | - |
| Mouse Expression Set 430 | 1420788_PM_at | - | - |
| Murine Genome U74v2 Set | 95320_PM_at | - | - |
| Murine Genome U74v2 Set | 95320_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420788_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420788_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420788_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420788_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 533816 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2360133 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002360133 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1254155 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1254155 | - | - |
| mouse-6 v1.1 (Illumina) | scl28445.5.1_99-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2360133 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl28445.5.1_99-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2360133 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2944755 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2944755 | - | - |
Citations (1)
Requirement for T-bet in the aberrant differentiation of unhelped memory CD8+ T cells.
- Authors
- Intlekofer AM, Takemoto N, Kao C, Banerjee A, Schambach F, Northrop JK, Shen H, Wherry EJ, Reiner SL
- Journal
- J Exp Med - Page No 2015 (Mon Sep 03 00:00:00 EDT 2007)
- Volume (Year)
- 204 (2007)
- Product Usage
- -

Klrg1
Genomic Map