Assay Details
| Gene Symbol: Grk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018869.3 | NP_061357.3 | 13 - 14 | 2239 | 58 | |
| GenBank mRNA | AF040746.1 | - | 13 - 14 | 1649 | 58 | |
| AK135076.1 | - | 13 - 14 | 2241 | 58 | ||
| AK161821.1 | - | 13 - 14 | 1719 | 58 | ||
| BC019379.1 | - | 13 - 14 | 1512 | 5116510 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Grk5
Genomic Map